Fentin fluoride

Supplier: Thermo Scientific Chemicals
Danger

39600-03 39600-06
CAAA39600-03EA 64.31 CAD
CAAA39600-03 CAAA39600-06
Fentin fluoride
Fentin fluoride

MDL: MFCD00015356


CAS No.: 379-52-2 Molecular Formula: (C6H5)3SnF Formula Weight: 368.99 Physical Form: Powder Melting Point: 281° UN No.: 3146 MDL No.: MFCD00015356

Formula: C₁₈H₁₅FSn
Melting Pt: >281 °C
Density: 1,5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00015356
CAS Number: 379-52-2
EINECS: 206-833-8
UN: 3146
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR