Diyttrium trisulfide ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Yttrium (III) sulphide, Yttrium sulfide, Yttrium (III) sulfide, Yttrium sulphide

39565-14 39565-03 39565-06
CAAAA39565-14EA 1845.44 CAD
CAAAA39565-14 CAAA39565-03 CAAA39565-06
Diyttrium trisulfide ≥99.9% (REO, rare earth oxide basis)
Diyttrium trisulphide

MDL: MFCD00058806 Notes: Decomposes in acid


Purity: 99.9% CAS No.: 12039-19-9 Molecular Formula: Y2S3 Formula Weight: 273.93 Physical Form: Powder Odor: Like rotten eggs UN No.: 3134 MDL No.: MFCD00058806

Formula: S₃Y₂
Storage Temperature: Ambient
MDL Number: MFCD00058806
CAS Number: 12039-19-9
EINECS: 234-887-2
UN: 3134
ADR: 4.3,II

Order Now

Specification Test Results

Form Powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR