Platinum iridium alloy (80:20 w%) ≥99.9% (metals basis), wire, annealed, Ø 0.2 mm (0.008 in)
Supplier: Thermo Scientific Chemicals
39526-BS
39526-BW
CAAA39526-BSEA
156.1
CAD
CAAA39526-BS
CAAA39526-BW
Platinum iridium alloy (80:20 w%) ≥99.9% (metals basis), wire, annealed, Ø 0.2 mm (0.008 in)
Platinum Iridium alloy (80:20 w%)
MDL: MFCD02091683
Purity: 99.9+%
Molecular Formula: Pt:Ir; 80:20 wt%
Physical Form: ≈0.7g/10cm, Temper: soft
Odor: Odorless
MDL No.: MFCD02091683
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...