Potassium methanesulfonate 99%
Supplier: Thermo Scientific Chemicals
39505-14
39505-22
39505-36
CAAA39505-14EA
99.7
CAD
CAAA39505-14
CAAA39505-22
CAAA39505-36
Potassium methanesulfonate 99%
Potassium methanesulphonate
MDL: MFCD00070544
Beilstein Registry No.: 3914899
Purity: 99%
CAS No.: 2386-56-3
Molecular Formula: CH3KO3S
Formula Weight: 134.19
Physical Form: Lump/powder
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Beilstein Reference: 3914899
MDL No.: MFCD00070544
Formula:
CH₃KO₃S Storage Temperature: Ambient |
MDL Number:
MFCD00070544 CAS Number: 2386-56-3 EINECS: 219-202-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...