Osmium(IV) oxide Os ≥ 83% (≥83% Os)
Supplier: Thermo Scientific Chemicals
|
Warning
|
39497-01
39497-03
39497-06
CAAA39497-01EA
210.14
CAD
CAAA39497-01
CAAA39497-03
CAAA39497-06
Osmium(IV) oxide Os ≥ 83% (≥83% Os)
Osmium (IV) oxide
MDL: MFCD00011150
CAS No.: 12036-02-1
Molecular Formula: OsO2
Formula Weight: 222.20
Physical Form: Solid
Density: 11.4
Appearance: Black
Odor: Chlorine-like
MDL No.: MFCD00011150
Formula:
O₂Os MW: 222.23 Density: 11.4 Storage Temperature: Ambient |
MDL Number:
MFCD00011150 CAS Number: 12036-02-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...