Palladium(II) trifluoroacetate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
39448-03
39448-06
CAAA39448-03EA
167.48
CAD
CAAA39448-03
CAAA39448-06
Palladium(II) trifluoroacetate 97%
Palladium (II) trifluoroacetate
MDL: MFCD00013204
Purity: 97%
CAS No.: 42196-31-6
Molecular Formula: Pd(O2CCF3)2
Formula Weight: 332.44
Physical Form: Powder
Sensitivity: Hygroscopic
MDL No.: MFCD00013204
Formula:
C₄F₆O₄Pd Melting Pt: 210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00013204 CAS Number: 42196-31-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...