Dipotassium tetraborate tetrahydrate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
39435-36
39435-A3
CAAA39435-36EA
75.37
CAD
CAAA39435-36
CAAA39435-A3
Dipotassium tetraborate tetrahydrate 99%
di-Potassium tetraborate tetrahydrate
MDL: MFCD00149924
Soluble in water
Purity: 99%
CAS No.: 12045-78-2
Molecular Formula: K2B4O7.4H2O
Formula Weight: 305.51 (233.44anhy)
Physical Form: Granular
Density: 1.74 (anhy)
Melting Point: dec.
Solubility: Soluble in water
Merck Reference: 14,7680
MDL No.: MFCD00149924
Formula:
B₄K₂O₇·4H₂O Melting Pt: Dec. Density: 1.74 (anhydr.) Storage Temperature: Ambient |
MDL Number:
MFCD00149924 CAS Number: 12045-78-2 EINECS: 215-575-5 Merck Index: 14,7680 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...