Terbium(III) bromide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
39425-03
39425-06
CAAA39425-03EA
241.74
CAD
CAAA39425-03
CAAA39425-06
Terbium(III) bromide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Terbium (III) bromide
MDL: MFCD00049571
Purity: 99.99%
CAS No.: 14456-47-4
Molecular Formula: TbBr3
Formula Weight: 398.65
Physical Form: Powder, Ampouled under argon
Sensitivity: Hygroscopic
Boiling Point: 1490°
MDL No.: MFCD00049571
Formula:
Br₃Tb Boiling Pt: 1490 °C Melting Pt: 827 °C Storage Temperature: Ambient |
MDL Number:
MFCD00049571 CAS Number: 14456-47-4 EINECS: 238-442-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...