Thulium(III) iodide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
39424-03
39424-06
CAAA39424-03EA
127.66
CAD
CAAA39424-03
CAAA39424-06
Thulium(III) iodide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Thulium (III) iodide
MDL: MFCD00050141
Purity: 99.99%
CAS No.: 13813-43-9
Molecular Formula: I3Tm
Formula Weight: 549.65
Physical Form: Flakes, Ampouled under argon
Sensitivity: Hygroscopic
Boiling Point: 1260°
MDL No.: MFCD00050141
Formula:
I₃Tm MW: 549.65 g/mol Boiling Pt: 1260 °C Melting Pt: 1015 °C Storage Temperature: Ambient |
MDL Number:
MFCD00050141 CAS Number: 13813-43-9 EINECS: 237-472-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...