Ammonium iron (III) sulfate dodecahydrate 98.5-102% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
39391-36
39391-A4
CAAA39391-36EA
145.52
CAD
CAAA39391-36
CAAA39391-A4
Ammonium iron (III) sulfate dodecahydrate 98.5-102% ACS
Ammonium iron (III) sulphate dodecahydrate
In dyeing and printing textiles, as analytical lab reagent
Purity: 98.5-102.0%
CAS No.: 7783-83-7
Molecular Formula: H4FeNO8S2.12H2O
Formula Weight: 482.18 (266.00anhy)
Physical Form: Broken Crystals
Density: 1.71
Appearance: Pale purple
Odor: Odorless
Melting Point: 39-41°
Solubility: Freely soluble in water. Insoluble in alcohol
Merck Reference: 14,521
MDL No.: MFCD00150004
In dyeing and printing textiles, as analytical lab reagent
Formula:
NH₄Fe(SO₄)₂·12H₂O Boiling Pt: 230 °C (1013 hPa) Melting Pt: 39…41 °C Density: 1.71 Storage Temperature: Ambient |
MDL Number:
MFCD00150004 CAS Number: 7783-83-7 EINECS: 233-382-4 Merck Index: 14,521 |
Specification Test Results
Insoluble matter | 0.01% |
Cl | 0.001% |
NO3 | 0.01% |
Ca | 0.01% |
MG | 0.005% |
K | 0.005% |
NA | 0.0002 |
Cu | 0.003% |
Ferrous iron | Pass test (limit about 0.001%) |
Zn | 0.003% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...