Praseodymium trichloride, ultra dry ≥99.99% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
39386-06
39386-14
CAAA39386-06EA
278.08
CAD
CAAA39386-06
CAAA39386-14
Praseodymium trichloride, ultra dry ≥99.99% (REO, rare earth oxide basis)
Praseodymium trichloride
MDL: MFCD00011176
Purity: 99.99%
CAS No.: 10361-79-2
Molecular Formula: PrCl3
Formula Weight: 247.27
Physical Form: Powder, Ampouled under argon
Density: 4.02
Melting Point: 786°
Boiling Point: 1700°
Merck Reference: 14,7712
MDL No.: MFCD00011176
Formula:
Cl₃Pr Boiling Pt: 1700 °C Melting Pt: 786 °C Density: 4.02 Storage Temperature: Ambient |
MDL Number:
MFCD00011176 CAS Number: 10361-79-2 EINECS: 233-794-4 Merck Index: 14,7712 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...