Praseodymium triiodide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
39385-03
39385-06
CAAA39385-03EA
152.79
CAD
CAAA39385-03
CAAA39385-06
Praseodymium triiodide, ultra dry ≥99.9% (REO, rare earth oxide basis)
Praseodymium triiodide
MDL: MFCD00049513
Purity: 99.9+%
CAS No.: 13813-23-5
Molecular Formula: I3Pr
Formula Weight: 521.62
Physical Form: Crystalline/Needles, Ampouled under argon
Sensitivity: Light Sensitive and Hygroscopic
MDL No.: MFCD00049513
Formula:
I₃Pr Melting Pt: 737 °C Storage Temperature: Ambient |
MDL Number:
MFCD00049513 CAS Number: 13813-23-5 EINECS: 237-466-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...