Dilithium zirconium trioxide ≥94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Lithium zirconate, Lithium zirconium oxide
39377-14
39377-22
CAAA39377-14EA
192.76
CAD
CAAA39377-14
CAAA39377-22
Dilithium zirconium trioxide ≥94%
di-Lithium zirconium trioxide
MDL: MFCD00016182
Purity: 94% min
CAS No.: 12031-83-3
Molecular Formula: Li2ZrO3
Formula Weight: 153.10
Physical Form: -80 Mesh Powder
Appearance: White to cream
Odor: Odorless
MDL No.: MFCD00016182
Formula:
Li₂O₃Zr MW: 153.1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00016182 CAS Number: 12031-83-3 EINECS: 234-760-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...