Lithium metavanadate ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Lithium vanadium trioxide, Lithium vanadium oxide, Lithium vanadate

39358-18 39358-30
CAAA39358-18EA 156.1 CAD
CAAA39358-18 CAAA39358-30
Lithium metavanadate ≥99.9% (metals basis)
Lithium metavanadate

MDL: MFCD00053997


Purity: 99.9% CAS No.: 15060-59-0 Molecular Formula: LiVO3 Formula Weight: 105.88 Physical Form: Powder UN No.: 3285 MDL No.: MFCD00053997

Formula: LiO₃V
MW: 105.88
Melting Pt: 605
Density: 2.74
Storage Temperature: Ambient
MDL Number: MFCD00053997
CAS Number: 15060-59-0
EINECS: 239-124-7
UN: 3285
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR