Lithium orthoarsenate 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
39350-06
39350-14
CAAA39350-06EA
80.58
CAD
CAAA39350-06
CAAA39350-14
Lithium orthoarsenate 99%
Lithium orthoarsenate
MDL: MFCD00053998
Soluble in dilute acetic acid. Insoluble in water
Purity: 99%
CAS No.: 13478-14-3
Molecular Formula: Li3AsO4
Formula Weight: 159.74
Physical Form: Powder
Density: 3.07
Solubility: Soluble in dilute acetic acid. Insoluble in water
UN No.: 1557
MDL No.: MFCD00053998
Formula:
AsLi₃O₄ Density: 3,07 Storage Temperature: Ambient |
MDL Number:
MFCD00053998 CAS Number: 13478-14-3 EINECS: 236-773-8 UN: 1557 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...