Lanthanum nitride ≥99.9% (REO, rare earth oxide basis)

Supplier: Thermo Scientific Chemicals
Warning

39342-06 39342-14
CAAA39342-06EA 200.66 CAD
CAAA39342-06 CAAA39342-14
Lanthanum nitride ≥99.9% (REO, rare earth oxide basis)
Lanthanum nitride

MDL: MFCD00049962


Purity: 99.9% CAS No.: 25764-10-7 Molecular Formula: LaN Formula Weight: 152.91 Physical Form: -60 Mesh Powder

Formula: LaN
MW: 152.91
Storage Temperature: Ambient
MDL Number: MFCD00049962
CAS Number: 25764-10-7
EINECS: 247-245-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR