Lithium stearate
Supplier: Thermo Scientific Chemicals
39335-30
39335-A1
CAAA39335-30EA
98.12
CAD
CAAA39335-30
CAAA39335-A1
Lithium stearate
Lithium stearate
MDL: MFCD00042032
CAS No.: 4485-12-5
Molecular Formula: CH3(CH2)16CO2Li
Formula Weight: 290.41
Physical Form: Powder
Melting Point: 220°
MDL No.: MFCD00042032
Formula:
C₁₈H₃₅LiO₂ Melting Pt: 220 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042032 CAS Number: 4485-12-5 EINECS: 224-772-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...