Lithium-2-ethylhexanoate

Supplier: Thermo Scientific Chemicals
Warning

39330-04 39330-09 39330-18
CAAA39330-04EA 47.24 CAD
CAAA39330-04 CAAA39330-09 CAAA39330-18
Lithium-2-ethylhexanoate
Lithium 2-ethylhexanoate

MDL: MFCD00058702


CAS No.: 15590-62-2 Molecular Formula: C8H15LiO2 Formula Weight: 150.15 Physical Form: Lump MDL No.: MFCD00058702

Formula: C₈H₁₅LiO₂
Storage Temperature: Ambient
MDL Number: MFCD00058702
CAS Number: 15590-62-2
EINECS: 239-657-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR