Iron(II) oxalate hydrate ≥96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Oxalic acid iron (II) salt, Ferrous oxalate, Iron (II) ethanedioate
39261-36
39261-A3
CAAA39261-36EA
104.12
CAD
CAAA39261-36
CAAA39261-A3
AA39261-22
Iron(II) oxalate hydrate ≥96%
Iron (II) oxalate
Used as a photographic developer, glass tint, decorative glass colorant, and pigment in plastics, paints, and lacquers.
Purity: 96%
CAS No.: 516-03-0
Molecular Formula: FeC2O4.xH2O (x≈2)
Formula Weight: 143.87(anhy)
Physical Form: Powder
Density: 2.28
Melting Point: 190° dec.
Solubility: Soluble in acids
UN No.: 3288
MDL No.: MFCD00012475
Formula:
FeC₂O₄·xH₂O MW: 143.87 Melting Pt: 190 °C (dec.) Density: 2.3 Storage Temperature: Ambient |
MDL Number:
MFCD00012475 CAS Number: 516-03-0 EINECS: 208-217-4 UN: 3288 ADR: 6.1 III Merck Index: 12,04099 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...