Aluminium lanthanum (III) oxide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals

39250-06 39250-14
CAAA39250-06EA 100.17 CAD
CAAA39250-06 CAAA39250-14
Aluminium lanthanum (III) oxide ≥99.9% (metals basis)
Aluminium lanthanum (III) oxide

MDL: MFCD00144550


Purity: 99.9% CAS No.: 12003-65-5 Molecular Formula: LaAlO3 Formula Weight: 213.88 Physical Form: Powder Odor: Odorless MDL No.: MFCD00144550

Formula: AlLaO₃
Storage Temperature: Ambient
MDL Number: MFCD00144550
CAS Number: 12003-65-5
EINECS: 234-433-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR