Hafnium(IV) telluride ≥99.5% (metals basis)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Hafnium telluride

39223-03 39223-06
CAAA39223-03EA 254.38 CAD
CAAA39223-03 CAAA39223-06
Hafnium(IV) telluride ≥99.5% (metals basis)
Hafnium (IV) telluride

MDL: MFCD00049947


Purity: 99.5% CAS No.: 39082-23-0 Molecular Formula: HfTe2 Formula Weight: 433.69 Physical Form: -325 Mesh Powder

Formula: HfTe₂
MW: 433.69
Storage Temperature: Ambient
MDL Number: MFCD00049947
CAS Number: 39082-23-0
EINECS: 254-280-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR