Caffeine ≥99.7%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, Methyltheobromine, 1,3,7-Trimethyl-2,6-dioxopurine, 2,6-Dihydroxy-1,3,7-trimethylpurine

39214-14 39214-22 39214-36
CAAA39214-14EA 60.04 CAD
CAAA39214-14 CAAA39214-22 CAAA39214-36
Caffeine ≥99.7%
Caffeine

MDL: MFCD00005758 Beilstein Registry No.: 17705


Purity: 99.7% CAS No.: 58-08-2 Molecular Formula: C8H10N4O2 Formula Weight: 194.19 Physical Form: Powder Density: 1.230 Appearance: White Melting Point: 234-236° Boiling Point: 178° subl. Beilstein Reference: 17705 Merck Reference: 14,1636 UN No.: 1544 MDL No.: MFCD00005758

Formula: C₈H₁₀N₄O₂
MW: 194.19 g/mol
Boiling Pt: 178 °C (subl.)
Melting Pt: 234…236 °C
Density: 1.230
Storage Temperature: Ambient
MDL Number: MFCD00005758
CAS Number: 58-08-2
EINECS: 200-362-1
UN: 1544
ADR: 6.1 III
Merck Index: 14,1636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=07e609e1c7a199176b9e5b938ef241d166d502f9&vid=1ac1145bba3574e2401439da90f37fee7fd7610b&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR