Thulium(III) bromide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
39186-03
39186-06
CAAA39186-03EA
195.92
CAD
CAAA39186-03
CAAA39186-06
Thulium(III) bromide, ultra dry ≥99.99% (REO, rare earth oxide basis)
Thulium (III) bromide
MDL: MFCD00050138
Purity: 99.99%
CAS No.: 14456-51-0
Molecular Formula: TmBr3
Formula Weight: 408.66
Physical Form: Powder, Ampouled under argon
Sensitivity: Hygroscopic
Boiling Point: 1440°
MDL No.: MFCD00050138
Formula:
Br₃Tm Boiling Pt: 1440 °C Melting Pt: 952 °C Storage Temperature: Ambient |
MDL Number:
MFCD00050138 CAS Number: 14456-51-0 EINECS: 238-444-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...