N-Methyl-2-pyrrolidone (NMP) ≥99.5% for spectrophotometry
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00003193
Beilstein Registry No.: 106420
Notes: Filtered through 0.2µ filters. Suitable for spectrophotometry, chromatography.
Fieser: 1,696 2,281 9,316
Purity: 99.5+%
CAS No.: 872-50-4
Molecular Formula: C5H9NO
Formula Weight: 99.13
Flash Point: 86°(187°F)
Physical Form: Liquid
Density: 1.033
Refractive Index: 1.4705
Sensitivity: Light Sensitive and Hygroscopic
Appearance: Colorless to pale yellow
Odor: Amine-like
Melting Point: -24°
Boiling Point: 204°
Beilstein Reference: 106420
Merck Reference: 14,6117
MDL No.: MFCD00003193
Formula:
C₅H₉NO Boiling Pt: 204 °C Melting Pt: –24 °C Density: 1.033 Flash Pt: 86 °C (187 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003193 CAS Number: 872-50-4 EINECS: 212-828-1 Merck Index: 14,6117 |
|
Specification Test Results
Evaporation residue | 6 ppm |
H2O | 0.0005 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...