1,4-Dioxane 99+% ACS

Supplier: Thermo Scientific Chemicals
Danger

39118-AP 39118-AY 39118-K2 39118-K7
CAAA39118-APEA 111.23 CAD
CAAA39118-AP CAAA39118-AY CAAA39118-K2 CAAA39118-K7
1,4-Dioxane 99+% ACS
1,4-Dioxane

MDL: MFCD00006571 Beilstein Registry No.: 102551 Fieser: 1,333


Purity: 99+% CAS No.: 123-91-1 Molecular Formula: C4H8O2 Formula Weight: 88.11 Flash Point: 12°(54°F) Physical Form: Liquid Density: 1.034 Refractive Index: 1.4220 Sensitivity: Hygroscopic Odor: Ether-like Melting Point: 11.8° Boiling Point: 100-102° Beilstein Reference: 102551 Merck Reference: 14,3300 UN No.: 1165 MDL No.: MFCD00006571

Formula: C₄H₈O₂
Boiling Pt: 100…102 °C
Melting Pt: 11.8 °C
Density: 1.034
Flash Pt: 12 °C (54 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006571
CAS Number: 123-91-1
EINECS: 204-661-8
UN: 1165
ADR: 3,II
Merck Index: 14,3300
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=df424642d4a7a96d2eebfb99cf9a2e6d128185d0&vid=37c3b8119f755f99624576b4e975000cbbb8349d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Freezing point not below 11.0°
Color (APHA) 20
Peroxide 0.005% max.
Evaporation residue 0.005%
Titratable acid 0.0016meq/g
Carbonyl (as HCHO) 0.01%
H2O 0.05%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR