Silicon ≥99.999% (metals basis), plate, Thickness: 6.35 mm (0.25 in)
Supplier: Thermo Scientific Chemicals
Warning
|
39112-FF
39112-FI
39112-GH
CAAA39112-FFEA
164.32
CAD
CAAA39112-FF
CAAA39112-FI
CAAA39112-GH
Silicon ≥99.999% (metals basis), plate, Thickness: 6.35 mm (0.25 in)
Silicon
MDL: MFCD00085311
Purity: 99.999%
CAS No.: 7440-21-3
Physical Form: ≈9.54g/25x25mm
Odor: Odorless
MDL No.: MFCD00085311
Formula:
Si Boiling Pt: 2355 °C (1013 hPa) Melting Pt: 1410 °C Density: 2.34 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00085311 CAS Number: 7440-21-3 EINECS: 231-130-8 Merck Index: 12,08635 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...