Silicon ≥99.999% (metals basis), plate, Thickness: 9.5 mm (0.37 in)

Supplier: Thermo Scientific Chemicals
Warning

39110-FF 39110-FI 39110-GH
CAAA39110-FFEA 316 CAD
CAAA39110-FF CAAA39110-FI CAAA39110-GH
Silicon ≥99.999% (metals basis), plate, Thickness: 9.5 mm (0.37 in)
Silicon

MDL: MFCD00085311


Purity: 99.999% CAS No.: 7440-21-3 Physical Form: ≈14.32g/25x25mm Odor: Odorless MDL No.: MFCD00085311

Formula: Si
Boiling Pt: 2355 °C (1013 hPa)
Melting Pt: 1410 °C
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
Merck Index: 12,08635

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR