Silicon ≥99.999% (metals basis), plate, Thickness: 12.5 mm (0.5 in)

Supplier: Thermo Scientific Chemicals
Warning

39109-FF 39109-FI 39109-GH
CAAA39109-FFEA 507.18 CAD
CAAA39109-FF CAAA39109-FI CAAA39109-GH
Silicon ≥99.999% (metals basis), plate, Thickness: 12.5 mm (0.5 in)
Silicon

MDL: MFCD00085311


Purity: 99.999% CAS No.: 7440-21-3 Physical Form: ≈19.09g/25x25mm Odor: Odorless MDL No.: MFCD00085311

Formula: Si
MW: 28.09 g/mol
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
Merck Index: 12,08635

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR