Iodine monobromide ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
39106-14
39106-22
CAAA39106-14EA
77.1
CAD
CAAA39106-14
CAAA39106-22
Iodine monobromide ≥98%
Iodine monobromide
MDL: MFCD00011353
Beilstein Registry No.: 3902973
Fieser: 1,501 2,224 17,152 18,193 20,200
Purity: 98% min
CAS No.: 7789-33-5
Molecular Formula: IBr
Formula Weight: 206.81
Flash Point: None
Physical Form: Crystalline
Density: 4.416
Sensitivity: Moisture and Light Sensitive
Appearance: Grey to black
Odor: Pungent
Melting Point: 50° subl
Boiling Point: 116° dec.
Beilstein Reference: 3902973
Merck Reference: 14,5016
UN No.: 3260
MDL No.: MFCD00011353
Formula:
IBr Boiling Pt: 116 °C (dec.) Melting Pt: 50 °C (subl.) Density: 4.416 Storage Temperature: Ambient |
MDL Number:
MFCD00011353 CAS Number: 7789-33-5 EINECS: 232-159-9 UN: 3260 ADR: 8,II Merck Index: 14,5016 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...