Silicon ≥99.999% (metals basis), rods, Ø 5.08cm (2.0 in)

Supplier: Thermo Scientific Chemicals
Warning

39103-DH 39103-DM
CAAA39103-DHEA 421.86 CAD
CAAA39103-DH CAAA39103-DM
Silicon ≥99.999% (metals basis), rods, Ø 5.08cm (2.0 in)
Silicon

MDL: MFCD00085311


Purity: 99.999% CAS No.: 7440-21-3 Physical Form: ≈47.18g/cm Odor: Odorless MDL No.: MFCD00085311

Formula: Si
Boiling Pt: 2355 °C (1013 hPa)
Melting Pt: 1410 °C
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
Merck Index: 12,08635

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR