Silicon ≥99.999% (metals basis), rods, Ø 2.5cm (0.98 in)

Supplier: Thermo Scientific Chemicals
Warning

39102-DH 39102-DM 39102-DO
CAAA39102-DHEA 311.26 CAD
CAAA39102-DH CAAA39102-DM CAAA39102-DO
Silicon ≥99.999% (metals basis), rods, Ø 2.5cm (0.98 in)
Silicon

MDL: MFCD00085311


Purity: 99.999% CAS No.: 7440-21-3 Physical Form: ≈11.43g/cm Odor: Odorless MDL No.: MFCD00085311

Formula: Si
Boiling Pt: 2355 °C (1013 hPa)
Melting Pt: 1410 °C
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
Merck Index: 12,08635

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR