Silicon ≥99.999% (metals basis), lumps 15 cm (5.9 in) & down

Supplier: Thermo Scientific Chemicals
Warning

39101-14 39101-22 39101-A1 39101-A4
CAAA39101-14EA 192.76 CAD
CAAA39101-14 CAAA39101-22 CAAA39101-A1 CAAA39101-A4
Silicon ≥99.999% (metals basis), lumps 15 cm (5.9 in) & down
Silicon

MDL: MFCD00085311


Purity: 99.999+% CAS No.: 7440-21-3 Odor: Odorless MDL No.: MFCD00085311

Formula: Si
Boiling Pt: 2355 °C (1013 hPa)
Melting Pt: 1410 °C
Density: 2.34 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00085311
CAS Number: 7440-21-3
EINECS: 231-130-8
Merck Index: 12,08635

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR