Titanium(III,IV) oxide ≥99.9% (metals basis)

Supplier: Thermo Scientific Chemicals

Synonyms: Titanium oxide

39000-14 39000-22
CAAA39000-14EA 186.44 CAD
CAAA39000-14 CAAA39000-22
Titanium(III,IV) oxide ≥99.9% (metals basis)
Titanium (III,IV) oxide

Used for optical coatings


Purity: 99.9% CAS No.: 12065-65-5 Molecular Formula: Ti3O5 Physical Form: 3-10mm Pieces Density: 4.20 Odor: Odorless MDL No.: MFCD00210650


Used for optical coatings

Formula: O₅Ti₃
MW: 223.64 g/mol
Melting Pt: 1800 °C
Density: 4,20
Storage Temperature: Ambient
MDL Number: MFCD00210650
CAS Number: 12065-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR