Glycerine, Ultrapure for HPLC
Supplier: Thermo Scientific Chemicals
|
Warning
|
MDL: MFCD00004722
Beilstein Registry No.: 635685
Common Applications: Useful as a cryoprotectant
Notes: Suitable for spectrophotometry, chromatography. Filtered through 0.2µ filters.
CAS No.: 56-81-5
Molecular Formula: HOCH2CH(OH)CH2OH
Formula Weight: 92.09
Flash Point: 160°(320°F)
Physical Form: Viscous liquid
Density: 1.26
Refractive Index: 1.474
Sensitivity: Hygroscopic
Appearance: Colorless
Odor: Odorless
Melting Point: 18°
Boiling Point: 182°/20mm
Beilstein Reference: 635685
Merck Reference: 14,4484
MDL No.: MFCD00004722
Formula:
HOCH₂CH(OH)CH₂OH Boiling Pt: 182 °C (20 mmHg) Melting Pt: 18 °C Density: 1.26 Flash Pt: 160 °C (320 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004722 CAS Number: 56-81-5 EINECS: 200-289-5 Merck Index: 14,4484 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...