N-Methyl-2-pyrrolidone (NMP) ≥99.5% for HPLC

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, 1-Methyl-2-pyrrolidone, NMP, Methyl Pyrrolidone

38986-K2 38986-K7 38986-M1
CAAA38986-K2EA 240.16 CAD
CAAA38986-K2 CAAA38986-K7 CAAA38986-M1
N-Methyl-2-pyrrolidone (NMP) ≥99.5% for HPLC
N-Methyl-2-pyrrolidone (NMP)

MDL: MFCD00003193 Beilstein Registry No.: 106420 Notes: Filtered through 0.2µ filters. Suitable for spectrophotometry, chromatography.


Purity: 99.5% CAS No.: 872-50-4 Molecular Formula: C5H9NO Formula Weight: 99.13 Flash Point: 86°(187°F) Physical Form: Liquid Density: 1.033 Refractive Index: 1.4705 Sensitivity: Light Sensitive and Hygroscopic Appearance: Colorless to pale yellow Odor: Amine-like Melting Point: -24° Boiling Point: 204° Beilstein Reference: 106420 Merck Reference: 14,6117 MDL No.: MFCD00003193

Formula: C₅H₉NO
Boiling Pt: 204 °C
Melting Pt: –24 °C
Density: 1.033
Flash Pt: 86 °C (187 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 14,6117
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR