Sulfuric acid 93-98%, Environmental Grade
Supplier: Thermo Scientific Chemicals
|
Danger
|
38751-AP
38751-K4
CAAA38751-APEA
297.04
CAD
CAAA38751-AP
CAAA38751-K4
Sulfuric acid 93-98%, Environmental Grade
Sulfuric acid
MDL: MFCD00064589
Notes: Certificate of Analysis included
CAS No.: 7664-93-9
Molecular Formula: H2SO4
Formula Weight: 98.08
Physical Form: Liquid, single sub-boiling quartz distillation
Density: 1.84
Appearance: Colorless to pale yellow
Odor: Acrid
Merck Reference: 14,8974
UN No.: 1830
MDL No.: MFCD00064589
Formula:
H₂SO₄ Boiling Pt: ∼335 °C (1013 hPa) Melting Pt: 3 °C Density: 1.84 Storage Temperature: Ambient |
MDL Number:
MFCD00064589 CAS Number: 7664-93-9 EINECS: 231-639-5 UN: 1830 ADR: 8,II Merck Index: 14,8974 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...