Hydrofluoric acid 47 - 51%, Environmental Grade Plus
Supplier: Thermo Scientific Chemicals
38746-AP
38746-AP
CAAA38746-APEA
2105
CAD
AA38746-AP
CAAA38746-AP
Hydrofluoric acid 47 - 51%, Environmental Grade Plus
Hydrofluoric acid
Formula:
HF MW: 20.01 g/mol Boiling Pt: 112 °C Density: 1.16 |
MDL Number:
MFCD00011346 CAS Number: 7664-39-3 UN: 1790 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...