Antimony ≥99.999% (metals basis), spherical shots 1-5 mm (0.04-0.20 in) low oxide
Supplier: Thermo Scientific Chemicals
|
Warning
|
38618-09
38618-18
CAAA38618-09EA
442.4
CAD
CAAA38618-09
CAAA38618-18
Antimony ≥99.999% (metals basis), spherical shots 1-5 mm (0.04-0.20 in) low oxide
Antimony
MDL: MFCD00134030
Purity: 99.999%
CAS No.: 7440-36-0
Physical Form: Ampouled under argon
Odor: Odorless
MDL No.: MFCD00134030
Formula:
Sb Boiling Pt: 1637 °C (1013 hPa) Melting Pt: 630 °C Density: 6.684 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134030 CAS Number: 7440-36-0 EINECS: 231-146-5 UN: 2871 ADR: 6.1,III Merck Index: 12,00733 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...