Barium fluoride, ultra dry ≥99.998% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Warning
|
38610-09
38610-18
CAAA38610-09EA
173.8
CAD
CAAA38610-09
CAAA38610-18
Barium fluoride, ultra dry ≥99.998% (metals basis)
Barium fluoride
MDL: MFCD00003450
Purity: 99.998%
CAS No.: 7787-32-8
Molecular Formula: BaF2
Formula Weight: 175.36
Physical Form: Powder, Ampouled under argon
Density: 4.83
Appearance: White
Odor: Odorless
Melting Point: 1353°
Boiling Point: 2260°
Merck Reference: 14,974
UN No.: 1564
MDL No.: MFCD00003450
Formula:
BaF₂ MW: 175.33 g/mol Boiling Pt: 2260 Melting Pt: 1270 Density: 4.83 Storage Temperature: Ambient |
MDL Number:
MFCD00003450 CAS Number: 7787-32-8 EINECS: 232-108-0 UN: 1564 ADR: 6.1 III Merck Index: 14,974 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...