Magnesium oxide substrate 10x10x1mm, polished one side 100 orientation
Supplier: Thermo Scientific Chemicals
Synonyms:
Magnesia usta
38503-KT
38503-KU
CAAA38503-KTEA
161.16
CAD
CAAA38503-KT
CAAA38503-KU
Magnesium oxide substrate 10x10x1mm, polished one side 100 orientation
Magnesium oxide
MDL: MFCD00011109
Molecular Formula: MgO
MDL No.: MFCD00011109
Formula:
MgO MW: 40.3 g/mol Boiling Pt: 3600 Melting Pt: 2800 Density: 3.58 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011109 CAS Number: 1309-48-4 EINECS: 215-171-9 Merck Index: 13,05700 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...