Chromium ≥99.995% (metals basis), pieces, Thickness: 2-3 mm (0.08-0.12 in)

Supplier: Thermo Scientific Chemicals

38494-18 38494-30
CAAA38494-18EA 165.9 CAD
CAAA38494-18 CAAA38494-30
Chromium ≥99.995% (metals basis), pieces, Thickness: 2-3 mm (0.08-0.12 in)
Chromium

MDL: MFCD00010944


Purity: 99.995% CAS No.: 7440-47-3 Physical Form: Broken plate, Large irregular pieces Odor: Odorless MDL No.: MFCD00010944

Formula: Cr
MW: 52 g/mol
Boiling Pt: 2469 °C (1013 hPa)
Melting Pt: 1840 °C
Density: 7.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010944
CAS Number: 7440-47-3
EINECS: 231-157-5
Merck Index: 12,02288

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR