Potassium bromide, crystal optic disc 25 mm x 2 mm, polished both sides

Supplier: Thermo Scientific Chemicals

38451-KT 38451-KT
CAAA38451-KTEA 152.94 CAD
CAAA38451-KT AA38451-KT
Potassium bromide, crystal optic disc 25 mm x 2 mm, polished both sides
Potassium bromide
Formula: KBr
MW: 119 g/mol
Boiling Pt: 1435 °C (1013 hPa)
Melting Pt: 730 °C
Density: 2.75 g/cm³ (20 °C)
MDL Number: MFCD00011358
CAS Number: 7758-02-3
Merck Index: 13,07701

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR