Strontium sulfate, REacton®, Reagent Grade

Supplier: Thermo Scientific Chemicals

38373-14 38373-22
CAAA38373-14EA 46.45 CAD
CAAA38373-14 CAAA38373-22
Strontium sulfate, REacton®, Reagent Grade
Strontium sulphate

MDL: MFCD00036274


CAS No.: 7759-02-6 Molecular Formula: SrSO4 Formula Weight: 183.68 Physical Form: Crystalline Density: 3.960 Odor: Odorless Merck Reference: 14,8848 MDL No.: MFCD00036274

Formula: O₄SSr
Melting Pt: 1600 °C
Density: 3.960
Storage Temperature: Ambient
MDL Number: MFCD00036274
CAS Number: 7759-02-6
EINECS: 231-850-2
Merck Index: 14,8848

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR