Zirconium ≥99.8% (metals basis excluding Hf), foil, annealed, Thickness: 0.025 mm

Supplier: Thermo Scientific Chemicals

38369-VR 38369-VS
CAAA38369-VREA 172.22 CAD
CAAA38369-VR CAAA38369-VS
Zirconium ≥99.8% (metals basis excluding Hf), foil, annealed, Thickness: 0.025 mm
Zirconium

MDL: MFCD00011303


Purity: 99.8% CAS No.: 7440-67-7 Physical Form: ≈8.3g/10x50cm, Temper: soft Odor: Odorless MDL No.: MFCD00011303

Formula: Zr
Storage Temperature: Ambient
MDL Number: MFCD00011303
CAS Number: 7440-67-7
EINECS: 231-176-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR