Cadmium sulfide ≥99.999% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
38342-18
38342-30
CAAA38342-18EA
287.56
CAD
CAAA38342-18
CAAA38342-30
Cadmium sulfide ≥99.999% (metals basis)
Cadmium sulphide
MDL: MFCD00010922
Purity: 99.999%
CAS No.: 1306-23-6
Molecular Formula: CdS
Formula Weight: 144.46
Physical Form: Powder/Granules
Density: 4.82
Appearance: Orange
Odor: Odorless
Melting Point: 980° subl.
Merck Reference: 14,1628
UN No.: 3077
MDL No.: MFCD00010922
Formula:
CdS Melting Pt: 980 °C (subl.) Density: 4.82 Storage Temperature: Ambient |
MDL Number:
MFCD00010922 CAS Number: 1306-23-6 EINECS: 215-147-8 UN: 3077 ADR: 9,III Merck Index: 14,1628 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...