Phenylmercury acetate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
MDL: MFCD00008691
Beilstein Registry No.: 3662930
Slightly soluble in water. Soluble in alcohol, benzene and glacial acetic acid
Purity: 98%
CAS No.: 62-38-4
Molecular Formula: C8H8HgO2
Formula Weight: 336.74
Physical Form: Powder
Density: 2.4
Appearance: White to yellow
Odor: Odorless
Melting Point: 146-153°
Solubility: Slightly soluble in water. Soluble in alcohol, benzene and glacial acetic acid
Beilstein Reference: 3662930
Merck Reference: 14,7300
UN No.: 1674
MDL No.: MFCD00008691
Formula:
C₈H₈HgO₂ Melting Pt: 146…153 °C Density: 2.4 Storage Temperature: Ambient |
MDL Number:
MFCD00008691 CAS Number: 62-38-4 EINECS: 200-532-5 UN: 1674 ADR: 6.1,II Merck Index: 14,7300 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...