Mercury(II) thiocyanate
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Mercury dithiocyanate, Mercuric thiocyanate, Mercury (II) rhodanide
37106-18
37106-30
CAAA37106-18EA
133.83
CAD
CAAA37106-18
CAAA37106-30
Mercury(II) thiocyanate
Mercury (II) thiocyanate
MDL: MFCD00011039
Beilstein Registry No.: 3561344
CAS No.: 592-85-8
Molecular Formula: C2HgN2S2
Formula Weight: 316.75
Physical Form: Powder
Density: 3.71
Sensitivity: Hygroscopic
Appearance: White to pale yellow
Odor: Odorless
Melting Point: ca 165° dec.
Beilstein Reference: 3561344
Merck Reference: 14,5890
UN No.: 1646
MDL No.: MFCD00011039
Formula:
C₂HgN₂S₂ MW: 316.76 g/mol Melting Pt: -38.9 (dec.) Density: 3.71 Flash Pt: 120 Storage Temperature: Ambient |
MDL Number:
MFCD00011039 CAS Number: 592-85-8 EINECS: 209-773-0 UN: 1646 ADR: 6.1 II Merck Index: 14,5890 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...