Lithium metaborate 98.0-102.0% ACS
Supplier: Thermo Scientific Chemicals
|
Warning
|
36760-18
36760-30
CAAA36760-18EA
156.26
CAD
CAAA36760-18
CAAA36760-30
Lithium metaborate 98.0-102.0% ACS
Lithium metaborate
MDL: MFCD00011089
Purity: 98.0-102.0%
CAS No.: 13453-69-5
Molecular Formula: BLiO2
Formula Weight: 49.75
Physical Form: Powder
Density: 1.397
Appearance: White
Odor: Odorless
Melting Point: 845°
MDL No.: MFCD00011089
Formula:
BLiO₂ Boiling Pt: 1719 °C (1013 hPa) Melting Pt: 845 °C Density: 1.397 Storage Temperature: Ambient |
MDL Number:
MFCD00011089 CAS Number: 13453-69-5 EINECS: 236-631-5 Merck Index: 13,05546 |
Specification Test Results
Bulk density | 0.25 g/ml |
Insoluble matter | 0.01% |
Loss on fusion at 950° | 2% |
Phosphorus compounds (as PO3) | 0.004% |
Si | 0.01% |
Al | 0.001% |
Ca | 0.01% |
Heavy metals (as Pb) | 0.001% |
Fe | 0.002% |
MG | 5ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...