Sodium metaborate tetrahydrate, tech.
Supplier: Thermo Scientific Chemicals
|
Warning
|
36736-A1
36736-A7
CAAA36736-A1EA
100.65
CAD
CAAA36736-A1
CAAA36736-A7
Sodium metaborate tetrahydrate, tech.
Sodium metaborate tetrahydrate
MDL: MFCD00149205
CAS No.: 10555-76-7
Molecular Formula: NaBO2.4H2O
Formula Weight: 137.86 (65.80anhy)
Physical Form: -8 Mesh Crystalline
Melting Point: ca 53° dec.
MDL No.: MFCD00149205
Formula:
BNaO₂·4H₂O MW: 137.86 g/mol Melting Pt: ∼ 53 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00149205 CAS Number: 10555-76-7 EINECS: 231-891-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...