Colophony (Rosin gum)
Supplier: Thermo Scientific Chemicals
|
Warning
|
36734-30
36734-A1
CAAA36734-30EA
59.72
CAD
CAAA36734-30
CAAA36734-A1
Colophony (Rosin gum)
Colophony (Rosin gum)
MDL: MFCD00132205
Soluble in alcohol, benzene, ether. Insoluble in water
CAS No.: 8050-09-7
Physical Form: Powder/solid
Solubility: Soluble in alcohol, benzene, ether. Insoluble in water
Merck Reference: 14,8266
MDL No.: MFCD00132205
Melting Pt:
75…81 °C Density: 1.1 g/cm³ (20 °C) Flash Pt: 188 °C Storage Temperature: Ambient |
MDL Number:
MFCD00132205 CAS Number: 8050-09-7 EINECS: 232-475-7 Merck Index: 14,8266 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...